missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXXC-type zinc finger protein 1
A DNA sequence encoding the Homo sapiens (Human) CXXC-type zinc finger protein 4, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Name | CXXC-type zinc finger protein 1 |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | CXXC4 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032871
|
enQuireBio™
QP8141-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052871
|
enQuireBio™
QP8141-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042871
|
enQuireBio™
QP8141-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062871
|
enQuireBio™
QP8141-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072871
|
enQuireBio™
QP8141-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082871
|
enQuireBio™
QP8141-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CXXC-type zinc finger protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
CXXC4 | |
Recombinant Protein | |
E. coli | |
MHHRNDSQRL GKAGCPPEPS LQMANTNFLS TLSPEHCRPL AGECMNKLKC GAAEAEIMNL PERVGTFSAI PALGGISLPP GVIVMTALHS PAAASAAVTD SAFQIANLAD CPQNHSSSSS SSSGGAGGAN PAKKKRKRCG VCVPCKRLIN CGVCSSCRNR KTGHQICKFR KCEELKKKPG TSLERTPVPS AEAFRWFF | |
Greater than 80% as determined by SDS-PAGE. |