missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CXXC-type zinc finger protein 1

A DNA sequence encoding the Homo sapiens (Human) CXXC-type zinc finger protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8141-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032871

  • 735.00 € / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CXXC-type zinc finger protein 1
Research Use Only
CXXC4
Recombinant Protein
E. coli
MHHRNDSQRL GKAGCPPEPS LQMANTNFLS TLSPEHCRPL AGECMNKLKC GAAEAEIMNL PERVGTFSAI PALGGISLPP GVIVMTALHS PAAASAAVTD SAFQIANLAD CPQNHSSSSS SSSGGAGGAN PAKKKRKRCG VCVPCKRLIN CGVCSSCRNR KTGHQICKFR KCEELKKKPG TSLERTPVPS AEAFRWFF
Greater than 80% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers