missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cyclophilin D Protein

A cDNA sequence encoding the Cyclophilin D was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10571-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984278

Quantity Price
1 1511.71 € / 1mg
3 1421.92 € / 1mg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

5481
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
PPID
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSHPSPQAKP SNPSNPRVFFDVDIGGERVG RIVLELFADI VPKTAENFRA LCTGEKGIGH TTGKPLHFKG CPFHRIIKKF MIQGGDFSNQ NGTGGESIYG EKFEDENFHY KHDREGLLSMANAGRNTNGS QFFITTVPTP HLDGKHVVFG QVIKGIGVAR ILENVEVKGEKPAKLCVIAE CGELKEGDDG GIFPKDGSGD SHPDFPEDAD IDLKDVDKIL LITEDLKNIG NTFFKSQNWE MAIKKYAEVL RYVDSSKAVI ETADRAKLQPIALSCVLNIG ACKLKMSNWQ GAIDSCLEAL ELDPSNTKAL YRRAQGWQGLKEYDQALADL KKAQGIAPED KAIQAELLKV KQKIKAQKDK EKAVYAKMFA
Cyclophilin D Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 210 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
Untagged
1 mg/ml solution containing 1x PBS pH 7.4 and 10% glycerol.
Videos
SDS
Documents
Certificates