missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cysteine-rich protein 1
A DNA sequence encoding the Homo sapiens (Human) Cysteine-rich protein 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001302.1 |
---|---|
Gene ID (Entrez) | 1396 |
Name | Cysteine-rich protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15938329
|
enQuireBio™
QP5868-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958329
|
enQuireBio™
QP5868-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948329
|
enQuireBio™
QP5868-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968329
|
enQuireBio™
QP5868-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978329
|
enQuireBio™
QP5868-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988329
|
enQuireBio™
QP5868-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001302.1 | |
Cysteine-rich protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1396 | |
Research Use Only | |
CRIP1 | |
Recombinant Protein | |
E. coli | |
PKCPKCNKEV YFAERVTSLG KDWHRPCLKC EKCGKTLTSG GHAEHEGKPY CNHPCYAAMF GPKGFGRGGA ESHTFK | |
Greater than 80% as determined by SDS-PAGE. |