missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CYTH1 Protein

A cDNA sequence encoding the CYTH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11589-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987728

  • 3401.86 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CYTH1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CYTH1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEEDDSY VPSDLTAEER QELENIRRRK QELLADIQRL KDEIAEVANE IENLGSTEER KNMQRNKQVA MGRKKFNMDP KKGIQFLIEN DLLKNTCEDI AQFLYKGEGL NKTAIGDYLG ERDEFNIQVL HAFVELHEFT DLNLVQALRQ FLWSFRLPGE AQKIDRMMEA FAQRYCQCNN GVFQSTDTCY VLSFAIIMLN TSLHNPNVKD KPTVERFIAM NRGINDGGDL PEELLRNLYE SIKNEPFKIP EDDGNDLTHT FFNPDREGWL LKLGGGRVKT WKRRWFILTD NCLYYFEYTT DKEPRGIIPL ENLSIREVED SKKPNCFELY IPDNKDQVIK ACKTEADGRV VEGNHTVYRI SAPTPEEKEE WIKCIKAAIS RDPFYEMLAA RKKKVSSTKR H
Videos
SDS
Documents
Certificates