missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CYTIP Protein

A cDNA sequence encoding the CYTIP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11592-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987738

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CYTIP Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CYTIP protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPPGSH MGSEFMSLQR LLQHSSNGNL ADFCAGPAYS SYSTLTGSLT MDDNRRIQML ADTVATLPRG RKQLALTRSS SLSDFSWSQR KLVTVEKQDN ETFGFEIQSY RPQNQNACSS EMFTLICKIQ EDSPAHCAGL QAGDVLANIN GVSTEGFTYK QVVDLIRSSG NLLTIETLNG TMILKRTELE AKLQVLKQTL KQKWVEYRSL QLQEHRLLHG DAANCPSLEN MDLDELSLFG PLPGPGPALV DRNRLSSESS CKSWLSSMTM DSEDGYQTCV SEDSSRGAFS RQTSTDDECF IPKEGDDFLR RSSSRRNRSI SNTSSGSMSP LWEGNLSSMF GTLPRKSRKG SVRKQLLKFI PGLHRAVEEE ESRF
Videos
SDS
Documents
Certificates