missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D-glucuronyl C5-epimerase Protein
A DNA sequence encoding the Homo sapiens (Human) D-glucuronyl C5-epimerase, was expressed in the hosts and tags indicated.
160.00 € - 2360.00 €
Specifications
Accession Number | NP_001311022.1 |
---|---|
Gene ID (Entrez) | 26035 |
Name | D-glucuronyl C5-epimerase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929869
|
enQuireBio™
QP6087-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949869
|
enQuireBio™
QP6087-EC-1MG |
1 mg |
2360.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939869
|
enQuireBio™
QP6087-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959869
|
enQuireBio™
QP6087-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969869
|
enQuireBio™
QP6087-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979869
|
enQuireBio™
QP6087-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001311022.1 | |
D-glucuronyl C5-epimerase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
26035 | |
Research Use Only | |
GLCE | |
Recombinant Protein | |
E. coli | |
NKCSSDKAIQ FPRRSSSGFR VDGFEKRAAA SESNNYMNHV AKQQSEEAFP QEQQKAPPVV GGFNSNVGSK VLGLKYEEID CLINDEHTIK GRREGNEVFL PFTWVEKYFD VYGKVVQYDG YDRFEFSHSY SKVYAQRAPY HPDGVFMSFE GYNVEVRDRV KCISGVEGVP LSTQWGPQGY FYPIQIAQYG LSHYSKNLTE KPPHIEVYET AEDRDKNKPN DWTVPKGCFM ANVADKSRFT NVKQFIAPET SEGVSLQLGN TKDFIISFDL KFLTNGSVSV VLETTEKNQL FTIHYVSNAQ LIAFKERDIY YGIGPRTSWS TVTRDLVTDL RKGVGLSNTK AVKPTKIMPK KVVRLIAKGK GFLDNITIST TAHMAAFFAA SDWLVRNQDE KGGWPIMVTR KLGEGFKSLE PGWYSAMAQG QAISTLVRAY LLTKDHIFLN SALRATAPYK FLSEQHGVKA VFMNKHDWYE EYPTTPSSFV LNGFMYSLIG LYDLKETAGE KLGKEARSLY ERGMESLKAM LPLYDTGSGT IYDLRHFMLG IAPNLARWDY HTTHINQLQL LSTIDESPVF KEFVKRWKSY LKGSRAKHN | |
Greater than 90% as determined by SDS-PAGE. |