missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCTN2 (1-401) Protein
A cDNA sequence encoding the DCTN2 (1-401) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11602-HIS-2ug
Additional Details : Weight : 0.01000kg
Specifications
DCTN2 (1-401) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADPKYA DLPGIARNEP DVYETSDLPE DDQAEFDAEE LTSTSVEHII VNPNAAYDKF KDKRVGTKGL DFSDRIGKTK RTGYESGEYE MLGEGLGVKE TPQQKYQRLL HEVQELTTEV EKIKTTVKES ATEEKLTPVL LAKQLAALKQ QLVASHLEKL LGPDAAINLT DPDGALAKRL LLQLEATKNS KGGSGGKTTG TPPDSSLVTY ELHSRPEQDK FSQAAKVAEL EKRLTELETA VRCDQDAQNP LSAGLQGACL METVELLQAK VSALDLAVLD QVEARLQSVL GKVNEIAKHK ASVEDADTQS KVHQLYETIQ RWSPIASTLP ELVQRLVTIK QLHEQAMQFG QLLTHLDTTQ QMIANSLKDN TTLLTQVQTT MRENLATVEG NFASIDERMK KLGK | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DCTN2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |