missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Death-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Death-associated protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8344-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001278892.1 | |
Death-associated protein 1 | |
Research Use Only | |
DAP1 | |
Recombinant Protein | |
E. coli | |
SSPPEGKLET KAGHPPAVKA GGMRIVQKHP HTGDTKEEKD KDDQEWESPS PPKPTVFISG VIARGDKDFP PAAAQVAHQK PHASMDKHPS PRTQHIQQPR K | |
Greater than 90% as determined by SDS-PAGE. |
1611 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |