missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DEFB118 Protein
A cDNA sequence encoding the DEFB118 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11618-1mg
Additional Details : Weight : 0.01000kg
Specifications
117285 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
DEFB118 | |
Human | |
His | |
DEFB118 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
DEFB118 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSYSGEKKC WNRSGHCRKQ CKDGEAVKDT CKNLRACCIP SNEDHRRVPA TSPTPLSDST PGIIDDILTV RFTTDYFEVS SKKDMVEESE AGRGTETSLP NVHHSS |