missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DHDDS Protein
A DNA sequence encoding the Homo sapiens (Human) DHDDS, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001230493.1 |
---|---|
Gene ID (Entrez) | 79947 |
Name | DHDDS Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958759
|
enQuireBio™
QP5932-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978759
|
enQuireBio™
QP5932-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968759
|
enQuireBio™
QP5932-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988759
|
enQuireBio™
QP5932-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998759
|
enQuireBio™
QP5932-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908769
|
enQuireBio™
QP5932-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001230493.1 | |
DHDDS Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
79947 | |
Research Use Only | |
DHDDS | |
Recombinant Protein | |
E. coli | |
MSWIKEGELS LWERFCANII KAGPMPKHIA FIMDGNRRYA KKCQVERQEG HSQGFNKLAE TLRWCLNLGI LEVTVYAFSI ENFKRSKSEV DGLMDLARQK FSRLMEEKEK LQKHGVCIRV LGDLHLLPLD LQELIAQAVQ ATKNYNKCFL NVCFAYTSRH EISNAVREMA WGVEQGLLDP SDISESLLDK CLYTNRSPHP DILIRTSGEV RLSDFLLWQT SHSCLVFQPV LWPEYTFWNL FEAILQFQMN HSVLQKARDM YAEERKRQQL ERDQATVTEQ LLREGLQASG DAQLRRTRLH KLSARREERV QGFLQALELK RADWLARLGT ASA | |
Greater than 80% as determined by SDS-PAGE. |