missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Diacylglycerol kinase alpha Protein
A DNA sequence encoding the Homo sapiens (Human) Diacylglycerol kinase alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5930-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001336.2 | |
Diacylglycerol kinase alpha Protein | |
Research Use Only | |
DGKA | |
Recombinant Protein | |
E. coli | |
MAKERGLISP SDFAQLQKYM EYSTKKVSDV LKLFEDGEMA KYVQGDAIGY EGFQQFLKIY LEVDNVPRHL SLALFQSFET GHCLNETNVT KDVVCLNDVS CYFSLLEGGR PEDKLEFTFK LYDTDRNGIL DSSEVDKIIL QMMRVAEYLD WDVSELRPIL QEMMKEIDYD GSGSVSQAEW VRAGATTVPL LVLLGLEMTL KDDGQHMWRP KRFPRPVYCN LCESSIGLGK QGLSCNLCKY TVHDQCAMKA LPCEVSTYAK SRKDIGVQSH VWVRGGCESG RCDRCQKKIR IYHSLTGLHC VWCHLEIHDD CLQAVGHECD CGLLRDHILP PSSIYPSVLA SGPDRKNSKT SQKTMDDLNL STSEALRIDP VPNTHPLLVF VNPKSGGKQG QRVLWKFQYI LNPRQVFNLL KDGPEIGLRL FKDVPDSRIL VCGGDGTVGW ILETIDKANL PVLPPVAVLP LGTGNDLARC LRWGGGYEGQ NLAKILKDLE MSKVVHMDRW SVEVIPQQTE EKSDPVPFQI INNYFSIGVD ASIAHRFHIM REKYPEKFNS RMKNKLWYFE FATSESIFST CKKLEESLTV EICGKPLDLS NLSLEGIAVL NIPSMHGGSN LWGDTRRPHG DIYGINQALG ATAKVITDPD ILKTCVPDLS DKRLEVVGLE GAIEMGQIYT KLKNAGRRLA KCSEITFHTT KTLPMQIDGE PWMQTPCTIK ITHKNQMPML MGPPPRSTNF FGFLS | |
Greater than 80% as determined by SDS-PAGE. |
1606 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |