missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA dC->dU-editing enzyme APOBEC-3A Protein
A DNA sequence encoding the Homo sapiens (Human) DNA dC->dU-editing enzyme APOBEC-3A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5664-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001180218.1 | |
DNA dC->dU-editing enzyme APOBEC-3A Protein | |
Research Use Only | |
APOBEC3A | |
Recombinant Protein | |
E. coli | |
MEASPASGPR HLMDPHIFTS NFNNGIGRHK TYLCYEVERL DNGTSVKMDQ HRGFLHNQAK NLLCGFYGRH AELRFLDLVP SLQLDPAQIY RVTWFISWSP CFSWGCAGEV RAFLQENTHV RLRIFAARIY DYDPLYKEAL QMLRDAGAQV SIMTYDEFKH CWDTFVDHQG CPFQPWDGLD EHSQALSGRL RAILQNQGN | |
Greater than 90% as determined by SDS-PAGE. |
100913187 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |