missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein complementing XP-G cells Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein complementing XP-G cells, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5994-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000114.2 | |
DNA repair protein complementing XP-G cells Protein | |
Research Use Only | |
ERCC5 | |
Recombinant Protein | |
E. coli | |
SFLWGKPDLD KIREFCQRYF GWNRTKTDES LFPVLKQLDA QQTQLRIDSF FRLAQQEKED AKRIKSQRLN RAVTCMLRKE KEAAASEIEA VSVAMEKEFE LLDKAKGKTQ KRGITNTLEE SSSLKRKRLS DSKGKNTCGG FLGETCLSES SDGSSSEDAE SSSLMNVQRR TAAKEPKTSA SDSQNSVKEA PVKNGGATTS SSSDSDDDGG KEKMVLVTAR SVFGKKRRKL RRARGRKRKT | |
Greater than 90% as determined by SDS-PAGE. |
2073 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |