missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DnaJ homolog subfamily B member 8 Protein
A DNA sequence encoding the Homo sapiens (Human) DnaJ homolog subfamily B member 8, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_699161.1 |
---|---|
Gene ID (Entrez) | 165721 |
Name | DnaJ homolog subfamily B member 8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051381
|
enQuireBio™
QP7884-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071381
|
enQuireBio™
QP7884-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061381
|
enQuireBio™
QP7884-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081381
|
enQuireBio™
QP7884-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091381
|
enQuireBio™
QP7884-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001391
|
enQuireBio™
QP7884-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_699161.1 | |
DnaJ homolog subfamily B member 8 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
165721 | |
Research Use Only | |
DNAJB8 | |
Recombinant Protein | |
E. coli | |
MANYYEVLGV QASASPEDIK KAYRKLALRW HPDKNPDNKE EAEKKFKLVS EAYEVLSDSK KRSLYDRAGC DSWRAGGGAS TPYHSPFDTG YTFRNPEDIF REFFGGLDPF SFEFWDSPFN SDRGGRGHGL RGAFSAGFGE FPAFMEAFSS FNMLGCSGGS HTTFSSTSFG GSSSGSSGFK SVMSSTEMIN GHKVTTKRIV ENGQERVEVE EDGQLKSVTV NGKEQLKWMD SK | |
Greater than 90% as determined by SDS-PAGE. |