missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAJB11 Protein
A cDNA sequence encoding the DNAJB11 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | DNAJB11 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987998
|
enQuireBio™
QP11675-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15908008
|
enQuireBio™
QP11675-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997998
|
enQuireBio™
QP11675-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DNAJB11 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DNAJB11 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGRDFYKILG VPRSASIKDI KKAYRKLALQ LHPDRNPDDP QAQEKFQDLG AAYEVLSDSE KRKQYDTYGE EGLKDGHQSS HGDIFSHFFG DFGFMFGGTP RQQDRNIPRG SDIIVDLEVT LEEVYAGNFV EVVRNKPVAR QAPGKRKCNC RQEMRTTQLG PGRFQMTQEV VCDECPNVKL VNEERTLEVE IEPGVRDGME YPFIGEGEPH VDGEPGDLRF RIKVVKHPIF ERRGDDLYTN VTISLVESLV GFEMDITHLD GHKVHISRDK ITRPGAKLWK KGEGLPNFDN NNIKGSLIIT FDVDFPKEQL TEEAREGIKQ LLKQGSVQKV YNGLQGY | |
Greater than 90.0% as determined by SDS-PAGE. |