missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dopa Decarboxylase Protein

A cDNA sequence encoding the Dopa Decarboxylase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11696-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988068

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

1644
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
DDC
Human
His
The Dopa decarboxylase protein solution (1 mg/ml) contains 20mM Tris-HCl, pH 8.0, 2mM DTT and 10% glycerol.
Dopa Decarboxylase Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH TRSMNASEFR RRGKEMVDYV ANYMEGIEGR QVYPDVEPGY LRPLIPAAAP QEPDTFEDII NDVEKIIMPG VTHWHSPYFF AYFPTASSYP AMLADMLCGA IGCIGFSWAA SPACTELETV MMDWLGKMLE LPKAFLNEKA GEGGGVIQGS ASEATLVALL AARTKVIHRL QAASPELTQA AIMEKLVAYS SDQAHSSVER AGLIGGVKLK AIPSDGNFAM RASALQEALE RDKAAGLIPF FMVATLGTTT CCSFDNLLEV GPICNKEDIW LHVDAAYAGS AFICPEFRHL LNGVEFADSF NFNPHKWLLV NFDCSAMWVK KRTDLTGAFR LDPTYLKHSH QDSGLITDYR HWQIPLGRRF RSLKMWFVFR MYGVKGLQAY IRKHVQLSHE FESLVRQDPR FEICVEVILG LVCFRLKGSN KVNEALLQRI NSAKKIHLVP CHLRDKFVLR FAICSRTVES AHVQRAWEHI KELAADVLRA ERE
Videos
SDS
Documents
Certificates