missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DPPA5 Protein
A cDNA sequence encoding the DPPA5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11700-10ug
Additional Details : Weight : 0.01000kg
Specifications
DPPA5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGTLPAR RHIPPWVKVP EDLKDPEVFQ VQTRLLKAIF GPDGSRIPYIEQVSKAMLEL KALESSDLTE VVVYGSYLYK LRTKWMLQSM AEWHRQRQER GMLKLAEAMN ALELGPWMK | |
Greater than 85.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DPPA5 protein solution (0.25 mg/ml) containing 20mM Phosphate buffer (pH 8.0), and 10% glycerol. |