missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP10 Protein
A cDNA sequence encoding the DUSP10 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | DUSP10 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15908118
|
enQuireBio™
QP11710-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998108
|
enQuireBio™
QP11710-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988108
|
enQuireBio™
QP11710-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DUSP10 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DUSP10 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMIIYPN DLAKKMTKCS KSHLPSQGPV IIDCRPFMEY NKSHIQGAVH INCADKISRR RLQQGKITVL DLISCREGKD SFKRIFSKEI IVYDENTNEP SRVMPSQPLH IVLESLKREG KEPLVLKGGL SSFKQNHENL CDNSLQLQEC REVGGGASAA SSLLPQPIPT TPDIENAELT PILPFLFLGN EQDAQDLDTM QRLNIGYVIN VTTHLPLYHY EKGLFNYKRL PATDSNKQNL RQYFEEAFEF IEEAHQCGKG LLIHCQAGVS RSATIVIAYL MKHTRMTMTD AYKFVKGKRP IISPNLNFMG QLLEFEEDLN NGVTPRILTP KLMGVETVV | |
Greater than 90% as determined by SDS-PAGE. |