missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DUSP10 Protein

A cDNA sequence encoding the DUSP10 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11710-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988108

  • 4534.09 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DUSP10 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DUSP10 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 50% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMIIYPN DLAKKMTKCS KSHLPSQGPV IIDCRPFMEY NKSHIQGAVH INCADKISRR RLQQGKITVL DLISCREGKD SFKRIFSKEI IVYDENTNEP SRVMPSQPLH IVLESLKREG KEPLVLKGGL SSFKQNHENL CDNSLQLQEC REVGGGASAA SSLLPQPIPT TPDIENAELT PILPFLFLGN EQDAQDLDTM QRLNIGYVIN VTTHLPLYHY EKGLFNYKRL PATDSNKQNL RQYFEEAFEF IEEAHQCGKG LLIHCQAGVS RSATIVIAYL MKHTRMTMTD AYKFVKGKRP IISPNLNFMG QLLEFEEDLN NGVTPRILTP KLMGVETVV
Videos
SDS
Documents
Certificates