missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 4, was expressed in the hosts and tags indicated.
202.00 € - 2645.00 €
Specifications
Accession Number | NP_001129115.1 |
---|---|
Gene ID (Entrez) | 51164 |
Name | Dynactin subunit 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049041
|
enQuireBio™
QP9937-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069041
|
enQuireBio™
QP9937-YE-1MG |
1 mg |
2645.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059041
|
enQuireBio™
QP9937-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079041
|
enQuireBio™
QP9937-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089041
|
enQuireBio™
QP9937-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099041
|
enQuireBio™
QP9937-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001129115.1 | |
Dynactin subunit 4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
51164 | |
Research Use Only | |
DCTN4 | |
Recombinant Protein | |
Yeast | |
ASLLQSDRVL YLVQGEKKVR APLSQLYFCR YCSELRSLEC VSHEVDSHYC PSCLENMPSA EAKLKKNRCA NCFDCPGCMH TLSTRATSIS TQLPDDPAKT TMKKAYYLAC GFCRWTSRDV GMADKSVASG GWQEPENPHT QRMNKLIEYY QQLAQKEKVE RDRKKLARRR NYMPLAFSDK YGLGTRLQRP RAGASISTLA GLSLKEGEDQ KEIKIEPAQA VDEVEPLPED YYTRPVNLTE VTTLQQRLLQ PDFQPVCASQ LYPRHKHLLI KRSLRCRKCE HNLSKPEFNP TSIKFKIQLV AVNYIPEVRI MSIPNLRYMK ESQVLLTLTN PVENLTHVTL FECEEGDPDD INSTAKVVVP PKELVLAGKD AAAEYDELAE PQDFQDDPDI IAFRKANKVG IFIKVTPQRE EGEVTVCFKM KHDFKNLAAP IRPIEESDQG TEVIWLTQHV ELSLGPLLP | |
Greater than 90% as determined by SDS-PAGE. |