missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dysbindin domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Dysbindin domain-containing protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8060-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001041686.1 | |
Dysbindin domain-containing protein 1 | |
Research Use Only | |
DBNDD2 | |
Recombinant Protein | |
E. coli | |
MDPNPRAALE RQQLRLRERQ KFFEDILQPE TEFVFPLSHL HLESQRPPIG SISSMEVNVD TLEQVELIDL GDPDAADVFL PCEDPPPTPQ SSGMDNHLEE LSLPVPTSDR TTSRTSSSSS SDSSTNLHSP NPSDDGADTP LAQSDEEEER GDGGAEPGAC S | |
Greater than 90% as determined by SDS-PAGE. |
55861 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |