missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E-Cadherin / CD324 Protein
A DNA sequence encoding the Homo sapiens (Human) E-Cadherin / CD324, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5803-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304113.1 | |
E-Cadherin / CD324 Protein | |
Research Use Only | |
CDH1 | |
Recombinant Protein | |
E. coli | |
DWVIPPISCP ENEKGPFPKN LVQIKSNKDK EGKVFYSITG QGADTPPVGV FIIERETGWL KVTEPLDRER IATYTLFSHA VSSNGNAVED PMEILITVTD QNDNKPEFTQ EVFKGSVMEG ALPGTSVMEV TATDADDDVN TYNAAIAYTI LSQDPELPDK NMFTINRNTG VISVVTTGLD RESFPTYTLV VQAADLQGEG LSTTATAVIT VTDTNDNPPI FNPTTYKGQV PENEANVVIT TLKVTDADAP NTPAWEAVYT ILNDDGGQFV VTTNPVNNDG ILKTAKGLDF EAKQQYILHV AVTNVVPFEV SLTTSTATVT VDVLDVNEAP IFVPPEKRVE VSEDFGVGQE ITSYTAQEPD TFMEQKITYR IWRDTANWLE INPDTGAIST RAELDREDFE HVKNSTYTAL IIATDNGSPV ATGTGTLLLI LSDVNDNAPI PEPRTIFFCE RNPKPQVINI IDADLPPNTS PFTAELTHGA SANWTIQYND PTQESIILKP KMALEVGDYK INLKLMDNQN KDQVTTLEVS VCDCEGAAGV CRKAQPVEAG LQI | |
Greater than 90% as determined by SDS-PAGE. |
999 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |