missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase SIAH1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase SIAH1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7820-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001006611.1 | |
E3 ubiquitin-protein ligase SIAH1 Protein | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6477 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
SIAH1 | |
Recombinant Protein | |
E. coli | |
MTGKATPPSL YSWRGVLFTC LPAARTRKRK EMSRQTATAL PTGTSKCPPS QRVPALTGTT ASNNDLASLF ECPVCFDYVL PPILQCQSGH LVCSNCRPKL TCCPTCRGPL GSIRNLAMEK VANSVLFPCK YASSGCEITL PHTEKADHEE LCEFRPYSCP CPGASCKWQG SLDAVMPHLM HQHKSITTLQ GEDIVFLATD INLPGAVDWV MMQSCFGFHF MLVLEKQEKY DGHQQFFAIV QLIGTRKQAE NFAYRLELNG HRRRLTWEAT PRSIHEGIAT AIMNSDCLVF DTSIAQLFAE NGNLGINVTI SMC |