missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase TRIM11 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase TRIM11, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_660215.1 |
---|---|
Gene ID (Entrez) | 81559 |
Name | E3 ubiquitin-protein ligase TRIM11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022101
|
enQuireBio™
QP8004-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042101
|
enQuireBio™
QP8004-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032101
|
enQuireBio™
QP8004-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052101
|
enQuireBio™
QP8004-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062101
|
enQuireBio™
QP8004-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072101
|
enQuireBio™
QP8004-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_660215.1 | |
E3 ubiquitin-protein ligase TRIM11 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
81559 | |
Research Use Only | |
TRIM11 | |
Recombinant Protein | |
E. coli | |
MELRTVCRVP GLVETLRRFR GDVTLDPDTA NPELILSEDR RSVQRGDLRQ ALPDSPERFD PGPCVLGQER FTSGRHYWEV EVGDRTSWAL GVCRENVNRK EKGELSAGNG FWILVFLGSY YNSSERALAP LRDPPRRVGI FLDYEAGHLS FYSATDGSLL FIFPEIPFSG TLRPLFSPLS SSPTPMTICR PKGGSGDTLA PQ | |
Greater than 90% as determined by SDS-PAGE. |