missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase ZNRF3 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase ZNRF3, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001193927.1 |
---|---|
Gene ID (Entrez) | 84133 |
Name | E3 ubiquitin-protein ligase ZNRF3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043461
|
enQuireBio™
QP8236-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063461
|
enQuireBio™
QP8236-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053461
|
enQuireBio™
QP8236-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073461
|
enQuireBio™
QP8236-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083461
|
enQuireBio™
QP8236-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093461
|
enQuireBio™
QP8236-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001193927.1 | |
E3 ubiquitin-protein ligase ZNRF3 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
84133 | |
Research Use Only | |
ZNRF3 | |
Recombinant Protein | |
E. coli | |
KETAFVEVVL FESSPSGDYT TYTTGLTGRF SRAGATLSAE GEIVQMHPLG LCNNNDEEDL YEYGWVGVVK LEQPELDPKP CLTVLGKAKR AVQRGATAVI FDVSENPEAI DQLNQGSEDP LKRPVVYVKG ADAIKLMNIV NKQKVARARI QHRPPRQPTE YFDM | |
Greater than 90% as determined by SDS-PAGE. |