missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EAF1 Protein

A cDNA sequence encoding the EAF1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11724-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988148

  • 6549.36 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EAF1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The EAF1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSDIIEQMS SSSGSSSSDS ESSSGSDDDS SSSGGEDNGP ASPPQPSHQQ PYNSRPAVAN GTSRPQGSNQ LMNTLRNDLQ LSESGSDSDD
Videos
SDS
Documents
Certificates