missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EGLN3 Protein

A cDNA sequence encoding the EGLN3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11761-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988278

Quantity Price
1 1511.71 € / 100µg
3 1421.92 € / 100µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

112399
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EGLN3
Human
His
The EGLN3 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 300mM NaCl, 5mM DTT, 2mM EDTA and 50% glycerol.
EGLN3 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMPLGHI MRLDLEKIAL EYIVPCLHEV GFCYLDNFLG EVVGDCVLER VKQLHCTGAL RDGQLAGPRA GVSKRHLRGD QITWIGGNEE GCEAISFLLS LIDRLVLYCG SRLGKYYVKE RSKAMVACYP GNGTGYVRHV DNPNGDGRCI TCIYYLNKNW DAKLHGGILR IFPEGKSFIA DVEPIFDRLL FFWSDRRNPH EVQPSYATRY AMTVWYFDAE ERAEAKKKFR NLTRKTESAL TED
Videos
SDS
Documents
Certificates