missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3E Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3E, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001559.1 |
---|---|
Gene ID (Entrez) | 3646 |
Name | EIF3E Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989029
|
enQuireBio™
QP5971-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909039
|
enQuireBio™
QP5971-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999029
|
enQuireBio™
QP5971-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919039
|
enQuireBio™
QP5971-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15929039
|
enQuireBio™
QP5971-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939039
|
enQuireBio™
QP5971-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001559.1 | |
EIF3E Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3646 | |
Research Use Only | |
EIF3E | |
Recombinant Protein | |
E. coli | |
AEYDLTTRIA HFLDRHLVFP LLEFLSVKEI YNEKELLQGK LDLLSDTNMV DFAMDVYKNL YSDDIPHALR EKRTTVVAQL KQLQAETEPI VKMFEDPETT RQMQSTRDGR MLFDYLADKH GFRQEYLDTL YRYAKFQYEC GNYSGAAEYL YFFRVLVPAT DRNALSSLWG KLASEILMQN WDAAMEDLTR LKETIDNNSV SSPLQSLQQR TWLIHWSLFV FFNHPKGRDN IIDLFLYQPQ YLNAIQTMCP HILRYLTTAV ITNKDVRKRR QVLKDLVKVI QQESYTYKDP ITEFVECLYV NFDFDGAQKK LRECESVLVN DFFLVACLED FIENARLFIF ETFCRIHQCI SINMLADKLN MTPEEAERWI VNLIRNARLD AKIDSKLGHV VMGNNAVSPY QQVIEKTKSL SFRSQMLAMN IEKKLNQNSR SEAPNWATQD SGFY | |
Greater than 80% as determined by SDS-PAGE. |