missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF4G1 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF4G1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5975-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004944.3 | |
EIF4G1 Protein | |
Research Use Only | |
EIF4G1 | |
Recombinant Protein | |
E. coli | |
IEEYLHLNDM KEAVQCVQEL ASPSLLFIFV RHGVESTLER SAIAREHMGQ LLHQLLCAGH LSTAQYYQGL YEILELAEDM EIDIPHVWLY LAELVTPILQ EGGVPMGELF REITKPLRPL GKAASLLLEI LGLLCKSMGP KKVGTLWREA GLSWKEFLPE GQDIGAFVAE QKVEYTLGEE SEAPGQRALP SEELNRQLEK LLKEGSSNQR VFDWIEANLS EQQIVSNTLV RALMTAVCYS AIIFETPLRV DVAVLKARAK LLQKYLCDEQ KELQALYALQ ALVVTLEQPP NLLRMFFDAL YDEDVVKEDA FYSWESSKDP AEQQGKGVAL KSVTAFFKWL REAEEESDHN | |
Greater than 90% as determined by SDS-PAGE. |
1981 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |