missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENPP1 Protein
A cDNA sequence encoding the ENPP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11790-2ug
Additional Details : Weight : 0.01000kg
Specifications
ENPP1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
HEK 293 | |
ASKPSCAKEV KSCKGRCFER TFGNCRCDAA CVELGNCCLD YQETCIEPEH IWTCNKFRCG EKRLTRSLCA CSDDCKDKGD CCINYSSVCQ GEKSWVEEPC ESINEPQCPA GFETPPTLLF SLDGFRAEYL HTWGGLLPVI SKLKKCGTYT KNMRPVYPTK TFPNHYSIVT GLYPESHGII DNKMYDPKMN ASFSLKSKEK FNPEWYKGEP IWVTAKYQGL KSGTFFWPGS DVEINGIFPD IYKMYNGSVP FEERILAVLQ WLQLPKDERP HFYTLYLEEP DSSGHSYGPV SSEVIKALQR VDGMVGMLMD GLKELNLHRC LNLILISDHG MEQGSCKKYI YLNKYLGDVK NIKVIYGPAA RLRPSDVPDK YYSFNYEGIA RNLSCREPNQ HFKPYLKHFL PKRLHFAKSD RIEPLTFYLD PQWQLALNPS ERKYCGSGFH GSDNVFSNMQ ALFVGYGPGF KHGIEADTFE NIEVYNLMCD LLNLTPAPNN GTHGSLNHLL KNPVYTPKHP KEVHPLVQCP FTRNPRDNLG CSCNPSILPI EDFQTQFNLT VAEEKIIKHE TLPYGRPRVL QKENTICLLS QHQFMSGYSQ DILMPLWTSY TVDRNDSFST EDFSNCLYQD FRIPLSPVHK CSFYKNNTKV SYGFLSPPQL NKNSSGIYSE ALLTTNIVPM YQSFQVIWRY FHDTLLRKYA EERNGVNVVS GPVFDFDYDG RCDSLENLRQ KRRVIRNQEI LIPTHFFIVL TSCKDTSQTP LHCENLDTLA FILPHRTDNS ESCVHGKHDS SWVEELLMLH RARITDVEHI TGLSFYQQRK EPVSDILKLK THLPTFSQED GPKLHHHHHH |