missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ephrin-A1 / EFNA1 Protein
A DNA sequence encoding the Homo sapiens (Human) Ephrin-A1 / EFNA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5963-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004419.2 | |
Ephrin-A1 / EFNA1 Protein | |
Research Use Only | |
EFNA1 | |
Recombinant Protein | |
E. coli | |
DRHTVFWNSS NPKFRNEDYT IHVQLNDYVD IICPHYEDHS VADAAMEQYI LYLVEHEEYQ LCQPQSKDQV RWQCNRPSAK HGPEKLSEKF QRFTPFTLGK EFKEGHSYYY ISKPIHQHED RCLRLKVTVS GKITHSPQAH DNPQEKRLAA DDPEVRVLHS IGHS | |
Greater than 90% as determined by SDS-PAGE. |
1942 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |