missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ephrin A4 Protein
A cDNA sequence encoding the Ephrin A4 was constructed and used to recombinantly synthesize the protein.
264.00 € - 715.00 €
Specifications
Gene ID (Entrez) | 1945 |
---|---|
Name | Ephrin A4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | EFNA4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968418
|
enQuireBio™
QP11800-2UG |
2 μg |
264.00 €
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978418
|
enQuireBio™
QP11800-5UG |
5 μg |
397.00 €
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958418
|
enQuireBio™
QP11800-10UG |
10 μg |
715.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1945 | |
Research Use Only | |
EFNA4 | |
Human | |
GST | |
Ephrin A4 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |
Ephrin A4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRLLPLLRTV LWAAFLGSPL RGGSSLRHVV YWNSSNPRLL RGDAVVELGL NDYLDIVCPH YEGPGPPEGP ETFALYMVDW PGYESCQAEG PRAYKRWVCS LPFGHVQFSE KIQRFTPFSL GFEFLPGETY YYISVPTPES SGQCLRLQVS VCCKERKSES AHPVGSPGES GTSGWRGGDT PSPLCLLLLL LLLILRLLRI L |