missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ETFA Protein

A DNA sequence encoding the Homo sapiens (Human) ETFA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6002-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959269

  • 735.00 € / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_000117.1
ETFA Protein
Research Use Only
ETFA
Recombinant Protein
E. coli
MFRAAAPGQL RRAASLLRFQ STLVIAEHAN DSLAPITLNT ITAATRLGGE VSCLVAGTKC DKVAQDLCKV AGIAKVLVAQ HDVYKGLLPE ELTPLILATQ KQFNYTHICA GASAFGKNLL PRVAAKLEVA PISDIIAIKS PDTFVRTIYA GNALCTVKCD EKVKVFSVRG TSFDAAATSG GSASSEKASS TSPVEISEWL DQKLTKSDRP ELTGAKVVVS GGRGLKSGEN FKLLYDLADQ LHAAVGASRA AVDAGFVPND MQVGQTGKIV APELYIAVGI SGAIQHLAGM KDSKTIVAIN KDPEAPIFQV ADYGIVADLF KVVPEMTEIL KKK
Greater than 80% as determined by SDS-PAGE.
2108
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers