missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ETNK2 Protein
A cDNA sequence encoding the ETNK2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11813-5ug
Additional Details : Weight : 0.01000kg
Specifications
ETNK2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAVPPSA PQPRASFHLR RHTPCPQCSW GMEEKAAASA SCREPPGPPR AAAVAYFGIS VDPDDILPGA LRLIQELRPH WKPEQVRTKR FTDGITNKLV ACYVEEDMQD CVLVRVYGER TELLVDRENE VRNFQLLRAH SCAPKLYCTF QNGLCYEYMQ GVALEPEHIR EPRLFRLIAL EMAKIHTIHA NGSLPKPILW HKMHNYFTLV KNEINPSLSA DVPKVEVLER ELAWLKEHLS QLESPVVFCH NDLLCKNIIY DSIKGHVRFI DYEYAGYNYQ AFDIGNHFNE FAGVNEVDYC LYPARETQLQ WLHYYLQAQK GMAVTPREVQ RLYVQVNKFA LASHFFWALW ALIQNQYSTI DFDFLRYAVI RFNQYFKVKP QASALEMPK | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ETNK2 protein solution (0.25 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |