missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC3 Protein
A cDNA sequence encoding the EXOSC3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11819-50ug
Additional Details : Weight : 0.01000kg
Specifications
EXOSC3 Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EXOSC3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 0.15M NaCl, 30% glycerol and 1mM DTT. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEPASV AAESLAGSRA RAARTVLGQV VLPGEELLLP EQEDAEGPGG AVERPLSLNA RACSRVRVVC GPGLRRCGDR LLVTKCGRLR HKEPGSGSGG GVYWVDSQQK RYVPVKGDHV IGIVTAKSGD IFKVDVGGSE PASLSYLSFE GATKRNRPNV QVGDLIYGQF VVANKDMEPE MVCIDSCGRA NGMGVIGQDG LLFKVTLGLI RKLLAPDCEI IQEVGKLYPL EIVFGMNGRI WVKAKTIQQT LILANILEAC EHMTSDQRKQ IFSRLAES |