missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EXOSC4 Protein

A cDNA sequence encoding the EXOSC4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11820-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988478

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

EXOSC4 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
EXOSC4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAGLELL SDQGYRVDGR RAGELRKIQA RMGVFAQADG SAYIEQGNTK ALAVVYGPHE IRGSRARALP DRALVNCQYS SATFSTGERK RRPHGDRKSC EMGLQLRQTF EAAILTQLHP RSQIDIYVQV LQADGGTYAA CVNAATLAVL DAGIPMRDFV CACSAGFVDG TALADLSHVE EAAGGPQLAL ALLPASGQIA LLEMDARLHE DHLERVLEAA AQAARDVHTL LDRVVRQHVR EASILLGD
Videos
SDS
Documents
Certificates