missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC5 Protein
A cDNA sequence encoding the EXOSC5 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | EXOSC5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15928488
|
enQuireBio™
QP11821-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918488
|
enQuireBio™
QP11821-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15908488
|
enQuireBio™
QP11821-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
EXOSC5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EXOSC5 solution (0.5 mg/1ml) contains PBS buffer (pH 7.4), 200mM Nacl, 2mM DTT and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMEEETHTDA KIRAENGTGS SPRGPGCSLR HFACEQNLLS RPDGSASFLQ GDTSVLAGVY GPAEVKVSKE IFNKATLEVI LRPKIGLPGV AEKSRERLIR NTCEAVVLGT LHPRTSITVV LQVVSDAGSL LACCLNAACM ALVDAGVPMR ALFCGVACAL DSDGTLVLDP TSKQEKEARA VLTFALDSVE RKLLMSSTKG LYSDTELQQC LAAAQAASQH VFRFYRESLQ RRYSKS | |
Greater than 90% as determined by SDS-PAGE. |