missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box / WD repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) F-box/WD repeat-containing protein 4, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_071322.1 |
---|---|
Gene ID (Entrez) | 6468 |
Name | F-box / WD repeat-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909439
|
enQuireBio™
QP6025-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929439
|
enQuireBio™
QP6025-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919439
|
enQuireBio™
QP6025-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939439
|
enQuireBio™
QP6025-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949439
|
enQuireBio™
QP6025-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959439
|
enQuireBio™
QP6025-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_071322.1 | |
F-box / WD repeat-containing protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6468 | |
Research Use Only | |
FBXW4 | |
Recombinant Protein | |
E. coli | |
MAAAAGEEEE EEEAARESAA RPAAGPALWR LPEELLLLIC SYLDMRALGR LAQVCRWLRR FTSCDLLWRR IARASLNSGF TRLGTDLMTS VPVKERVKVS QNWRLGRCRE GILLKWRCSQ MPWMQLEDDS LYISQANFIL AYQFRPDGAS LNRRPLGVFA GHDEDVCHFV LANSHIVSAG GDGKIGIHKI HSTFTVKYSA HEQEVNCVDC KGGIIVSGSR DRTAKVWPLA SGRLGQCLHT IQTEDRVWSI AISPLLSSFV TGTACCGHFS PLRIWDLNSG QLMTHLGSDF PPGAGVLDVM YESPFTLLSC GYDTYVRYWD LRTSVRKCVM EWEEPHDSTL YCLQTDGNHL LATGSSYYGV VRLWDRRQRA CLHAFPLTST PLSSPVYCLR LTTKHLYAAL SYNLHVLDFQ NP | |
Greater than 90% as determined by SDS-PAGE. |