missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP3 Protein
A cDNA sequence encoding the FABP3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | FABP3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948528
|
enQuireBio™
QP11835-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938528
|
enQuireBio™
QP11835-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928528
|
enQuireBio™
QP11835-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FABP3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
50mM phosphate borate buffer pH 8. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVDAFLGTWK LVDSKNFDDY MKSLGVGFAT RQVASMTKPT TIIEKNGDIL TLKTHSTFKN TEISFKLGVE FDETTADDRK VKSIVTLDGG KLVHLQKWDG QETTLVRELI DGKLILTLTH GTAVCTRTYE KEA | |
Greater than 90.0% as determined by SDS-PAGE. |