missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP9 Protein

A cDNA sequence encoding the FABP9 was constructed and used to recombinantly synthesize the protein.

131.00 € - 3915.00 €

Specifications

Name FABP9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15908548
View Documents Promotion Details
enQuireBio™
QP11838-5UG
5 μg
131.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15998538
View Documents Promotion Details
enQuireBio™
QP11838-20UG
20 μg
203.00 €
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15988538
View Documents Promotion Details
enQuireBio™
QP11838-1MG
1 mg
3915.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

FABP9 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FABP9 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMVEPFL GTWKLVSSEN FEDYMKELGV NFAARNMAGL VKPTVTISVD GKMMTIRTES SFQDTKISFK LGEEFDETTA DNRKVKSTIT LENGSMIHVQ KWLGKETTIK RKIVDEKMVV ECKMNNIVST RIYEKV
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications