missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM50A Protein
A cDNA sequence encoding the FAM50A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11848-1mg
Additional Details : Weight : 0.01000kg
Specifications
FAM50A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTTKKRKL GKNPDVDTSF LPDRDREEEE NRLREELRQE WEAKQEKIKS EEIEITFSYW DGSGHRRTVK MRKGNTMQQF LQKALEILRK DFSELRSAGV EQLMYIKEDL IIPHHHSFYD FIVTKARGKS GPLFNFDVHD DVRLLSDATV EKDESHAGKV VLRSWYEKNK HIFPASRWEP YDPEKKWDKY TIR | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FAM50A protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |