missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FARSB Protein

A cDNA sequence encoding the FARSB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11852-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928588

  • 120.00 € / 5µg
Estimated Shipment: 20-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FARSB Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FARSB protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPTVSVK RDLLFQALGR TYTDEEFDEL CFEFGLELDE ITSEKEIISK EQGNVKAAGA SDVVLYKIDV PANRYDLLCL EGLVRGLQVF KERIKAPVYK RVMPDGKIQK LIITEETAKI RPFAVAAVLR NIKFTKDRYD SFIELQEKLH QNICRKRALV AIGTHDLDTL SGPFTYTAKR PSDIKFKPLN KTKEYTACEL MNIYKTDNHL KHYLHIIENK PLYPVIYDSN GVVLSMPPII NGDHSRITVN TRNIFIECTG TDFTKAKIVL DIIVTMFSEY CENQFTVEAA EVVFPNGKSH TFPELAYRKE MVRADLINKK VGIRETPENL AKLLTRMYLK SEVIGDGNQI EIEIPPTRAD IIHACDIVED AAIAYGYNNI QMTLPKTYTI ANQFPLNKLT ELLRHDMAAA GFTEALTFAL CSQEDIADKL GVDISATKAV HISNPKTAEF QVARTTLLPG LLKTIAANRK MPLPLKLFEI SDIVIKDSNT DVGAKNYRHL CAVYYNKNPG FEIIHGLLDR IMQLLDVPPG EDKGGYVIKA SEGPAFFPGR CAEIFARGQS VGKLGVLHPD VITKFELTMP CSSLEINVGP FL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers