missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fas Ligand / FASLG / CD95L Protein
A DNA sequence encoding the Homo sapiens (Human) Fas Ligand / FASLG / CD95L, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_000630.1 |
---|---|
Gene ID (Entrez) | 356 |
Name | Fas Ligand / FASLG / CD95L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969409
|
enQuireBio™
QP6021-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989409
|
enQuireBio™
QP6021-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979409
|
enQuireBio™
QP6021-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15999409
|
enQuireBio™
QP6021-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15909419
|
enQuireBio™
QP6021-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919419
|
enQuireBio™
QP6021-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000630.1 | |
Fas Ligand / FASLG / CD95L Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
356 | |
Research Use Only | |
FASLG | |
Recombinant Protein | |
E. coli | |
QIGHPSPPPE KKELRKVAHL TGKSNSRSMP LEWEDTYGIV LLSGVKYKKG GLVINETGLY FVYSKVYFRG QSCNNLPLSH KVYMRNSKYP QDLVMMEGKM MSYCTTGQMW ARSSYLGAVF NLTSADHLYV NVSELSLVNF EESQTFFGLY KL | |
Greater than 90% as determined by SDS-PAGE. |