missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FBXO6 Protein
A cDNA sequence encoding the FBXO6 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | FBXO6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948598
|
enQuireBio™
QP11856-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938598
|
enQuireBio™
QP11856-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928598
|
enQuireBio™
QP11856-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FBXO6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FBXO6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDAPHSK AALDSINELP ENILLELFTH VPARQLLLNC RLVCSLWRDL IDLMTLWKRK CLREGFITKD WDQPVADWKI FYFLRSLHRN LLRNPCAEED MFAWQIDFNG GDRWKVESLP GAHGTDFPDP KVKKYFVTSY EMCLKSQLVD LVAEGYWEEL LDTFRPDIVV KDWFAARADC GCTYQLKVQL ASADYFVLAS FEPPPVTIQQ WNNATWTEVS YTFSDYPRGV RYILFQHGGR DTQYWAGWYG PRVTNSSIVV SPKMTRNQAS SEAQPGQKHG QEEAAQSPYR AVVQIF | |
Greater than 90% as determined by SDS-PAGE. |