missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fc receptor-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Fc receptor-like protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7876-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_112572.1 | |
Fc receptor-like protein 1 | |
Research Use Only | |
FCRL4 | |
Recombinant Protein | |
E. coli | |
AHKPVISVHP PWTTFFKGER VTLTCNGFQF YATEKTTWYH RHYWGEKLTL TPGNTLEVRE SGLYRCQARG SPRSNPVRLL FSSDSLILQA PYSVFEGDTL VLRCHRRRKE KLTAVKYTWN GNILSISNKS WDLLIPQASS NNNGNYRCIG YGDENDVFRS NFKIIKIQEL FPHPELKATD SQPTEGNSVN LSCETQLPPE RSDTPLHFNF FRDGEVILSD WSTYPELQLP TVWRENSGSY WCGAETVRGN IHKHSPSLQI HVQRIPVSGV LLETQPSGGQ AVEGEMLVLV CSVAEGTGDT TFSWHREDMQ ESLGRKTQRS LRAELELPAI RQSHAGGYYC TADNSYGPVQ SMVLNVTVRE TPGNRDGL | |
Greater than 90% as determined by SDS-PAGE. |
83417 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |