missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCGR1A Protein
A cDNA sequence encoding the FCGR1A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11858-100ug
Additional Details : Weight : 0.01000kg
Specifications
2209 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMQVDTT KAVITLQPPW VSVFQEETVT LHCEVLHLPG SSSTQWFLNG TATQTSTPSY RITSASVNDS GEYRCQRGLS GRSDPIQLEI HRGWLLLQVS SRVFTEGEPL ALRCHAWKDK LVYNVLYYRN GKAFKFFHWN SNLTILKTNI SHNGTYHCSG MGKHRYTSAG ISVTVKELFP APVLNASVTS PLLEGNLVTL SCETKLLLQR PGLQLYFSFY MGSKTLRGRN TSSEYQILTA RREDSGLYWC EAATEDGNVL KRSPELELQV LGLQLPTPVW FH | |
Greater than 85% as determined by SDS-PAGE. |
FCGR1A Protein | |
Research Use Only | |
FCGR1A | |
Human | |
His | |
The FCGR1A solution contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |