missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF6 Protein

A cDNA sequence encoding the FGF6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11872-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988648

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

FGF6 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
FGF-6 protein was lyophilized from a 0.2microm filtered solution in 10mM sodium phosphate and 50mM sodium chloride pH 7.5.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
Videos
SDS
Documents
Certificates