missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR2 Protein
A cDNA sequence encoding the FGFR2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 2263 |
---|---|
Name | FGFR2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | FGFR2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984388
|
enQuireBio™
QP10605-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964388
|
enQuireBio™
QP10605-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974388
|
enQuireBio™
QP10605-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
2263 | |
Research Use Only | |
FGFR2 | |
Recombinant Protein | |
Baculovirus | |
MVSWGRFICL VVVTMATLSL ARPSFSLVED TTLEPEEPPT KYQISQPEVY VAAPGESLEV RCLLKDAAVI SWTKDGVHLG PNNRTVLIGE YLQIKGATPR DSGLYACTAS RTVDSETWYF MVNVTDAISS GDDEDDTDGA EDFVSENSNN KRAPYWTNTE KMEKRLHAVP AANTVKFRCP AGGNPMPTMR WLKNGKEFKQ EHRIGGYKVR NQHWSLIMES VVPSDKGNYT CVVENEYGSI NHTYHLDVVE RSPHRPILQA GLPANASTVV GGDVEFVCKV YSDAQPHIQW IKHVEKNGSK YGPDGLPYLK VLKAAGVNTT DKEIEVLYIR NVTFEDAGEY TCLAGNSIGI SFHSAWLTVL PAPGREKEIT ASPDYLEIAI YCIGVFLIAC MVVTVILCRM KNTTKKPDFS SQPAVHKLTK RIPLRRQVTV SAESSSSMNS NTPLVRITTR LSSTADTPML AGVSEYELPE DPKWEFPRDK LTLGKPLGEG CFGQVVMAEA VGIDKDKPKE AVTVAVKMLK DDATEKDLSD LVSEMEMMKM IGKHKNIINL LGACTQDGPL YVIVEYASKG NLREYLRARR PPGMEYSYDI NRVPEEQMTF KDLVSCTYQL ARGMEYLASQ KCIHRDLAAR NVLVTENNVM KIADFGLARD INNIDYYKKT TNGRLPVKWM APEALFDRVY THQSDVWSFG VLMWEIFTLG GSPYPGIPVE ELFKLLKEGH RMDKPANCTN ELYMMMRDCW HAVPSQRPTF KQLVEDLDRI LTLTTNEEYL DLSQPLEQYS PSYPDTRSSC SSGDDSVFSP DPMPYEPCLP QYPHINGSVK T | |
Greater than 90.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
FGFR2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Determined by its ability to inhibit human FGF acidic-dependent proliferation on R1 cells. The ED50 for this effect is typically at 15.0-30.0 ng/ml. | |
Human | |
Untagged | |
CD332 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |