missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FHL2 Protein
A cDNA sequence encoding the FHL2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | FHL2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15998668
|
enQuireBio™
QP11879-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988668
|
enQuireBio™
QP11879-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978668
|
enQuireBio™
QP11879-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FHL2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FHL2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTERFDC HHCNESLFGK KYILREESPY CVVCFETLFA NTCEECGKPI GCDCKDLSYK DRHWHEACFH CSQCRNSLVD KPFAAKEDQL LCTDCYSNEY SSKCQECKKT IMPGTRKMEY KGSSWHETCF ICHRCQQPIG TKSFIPKDNQ NFCVPCYEKQ HAMQCVQCKK PITTGGVTYR EQPWHKECFV CTACRKQLSG QRFTARDDFA YCLNCFCDLY AKKCAGCTNP ISGLGGTKYI SFEERQWHND CFNCKKCSLS LVGRGFLTER DDILCPDCGK DI | |
Greater than 90.0% as determined by SDS-PAGE. |